Quanti-Cult Plus™ Staphylococcus epidermidis ATCC™ 12228™
Catalog # OXOIR4716500
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Staphylococcus epidermidis ATCC™ 12228™
Specifications
About this item
Quanti-Cult Plus™ Staphylococcus epidermidis ATCC™ 12228™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Packaging: Includes 10 preserved organism vials and 10 rehydration vials.