Quanti-Cult Plus™ Staphylococcus aureus ATCC™ 29737™
OXOIR4717020
:
- Cdt:1 KIT
- Désignation:Staphylococcus aureus ATCC™ 29737™
Quanti-Cult Plus™ Staphylococcus aureus subsp. aureus ATCC™ 29737™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
: Includes 10 preserved organism vials and 10 rehydration vials.