Culti-Loops™ Sphingobacterium multivorum ATCC™ 35656™
Sphingobacterium multivorum ATCC™ 35656™, 5
Catalog # OXOIR4602004
Supplier: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Sphingobacterium multivorum</i> ATCC™ 35656™
Catalog # OXOIR4602004
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Jump to:
Product Details & Documents
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
- Cdt:5
- Désignation:Sphingobacterium multivorum ATCC™ 35656™