Quanti-Cult Plus™ Escherichia coli ATCC™ 25922™
OXOIR4717050
:
- Cdt:100 Tests
- Désignation:Escherichia coli ATCC™ 25922™
Quanti-Cult Plus™ Escherichia coli ATCC™ 25922™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
- 10 tests/reconstituted vial; 100 tests/kit
- Ready for rehydration
- For use in quality control procedures
: Includes 10 preserved organism vials and 10 rehydration vials.