Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 19404™
N° de catalogue OXOIR4711700
Fournisseur: THERMO FISHER DIAGNOSTICS
Numéro CAS:
Spécifications
- Cdt:100 Tests
- Désignation:Clostridium sporogenes ATCC™ 19404™
Spécifications
À propos de cet article
Quanti-Cult Plus™ Clostridium sporogenes ATCC™ 19404™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
- Ready for rehydration
- For use in quality control procedures
Emballage: Includes 10 preserved organism vials and 10 rehydration vials.