Quanti-Cult Plus™ Candida albicans ATCC™ 10231™
Catalog # OXOIR4711503
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Cdt:1
- Désignation:Candida albicans ATCC™ 10231™
- Seq:02
Specifications
About this item
Quanti-Cult Plus™ Candida albicans ATCC™ 10231™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
- No serial dilution required
- Delivers <100 colony forming units (CFUs) in a single 0,1 ml inoculum
- Ready for rehydration
- For use in quality control procedures
Packaging: Yields 10 tests/reconstituted vial, 100 tests/kit.