Culti-Loops™ Enterococcus faecium ATCC™ 51559™
Enterococcus faecium ATCC™ 51559™, 5
N° de catalogue OXOIR4609383
Fournisseur: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Enterococcus faecium</i> ATCC™ 51559™
N° de catalogue OXOIR4609383
Fournisseur: THERMO FISHER DIAGNOSTICS
Numéro CAS:
Aller à :
Product Details & Documents
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Spécifications
Spécifications
- Cdt:5
- Désignation:Enterococcus faecium ATCC™ 51559™