Anti-RNF210 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
N° de catalogue BOSSBS-8494R-A750
Fournisseur: Bioss
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:RNF210
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 750
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat
- Western blot:Yes
- Épitope:101-200/287
- Formulaire:Liquid
- ID de gène:342931
- Synonymes antigène:RFPL4A|RFPLA_HUMAN|RNF210|Ret finger protein-like 4A|RFPL4|Ret finger protein like 4|RING finger protein 210
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:32 kDa
- Température de stockage:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human RFPL4A/RNF210
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
RNF210.
Type: Primary
Antigen: RNF210
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/287
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat