Anti-5HT3C Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5328R-A750
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:5HT3C
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 750
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Épitope:101-200/447
- Formulaire:Liquid
- ID de gène:285242
- Synonymes antigène:5HT3E|serotonin receptor 3 subunit E|Serotonin receptor 3E|HTR3|5HT3R|5-hydroxytryptamine receptor 3E|SEROTONIN 5-HT-3E RECEPTOR|5-hydroxytryptamine receptor 3 subunit E|5-HT3c1|5-hydroxytryptamine serotonin receptor 3, family member E|5-HT-3|5-HT3R|5-HT3E|5-HT3 receptor subunit E splice variant HTR3Ea|5HT3c1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:47 kDa
- Température de stockage:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human 5HT3C
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Specifications
About this item
HTR3C belongs to the ligand-gated ion channel receptor superfamily. HTR3E is a subunit E of the type 3 receptor for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarising responses in neurons after activation. Genes encoding subunits C, D and E form a cluster on chromosome 3. An alternative splice variant has been described but its full length sequence has not been determined.
Type: Primary
Antigen: 5HT3C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/447
Host: Rabbit
Isotype: IgG
Reactivity: