Order Entry
Belgium
ContactUsLinkComponent
Anti-DHPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-DHPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
N° de catalogue BOSSBS-8260R-A750
Fournisseur:  Bioss
Anti-DHPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
N° de catalogue BOSSBS-8260R-A750
Fournisseur:  Bioss

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    DHPS1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Alexa Fluor® 750
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Mouse
  • Western blot:
    Yes
  • Épitope:
    1-100/313
  • Formulaire:
    Liquid
  • ID de gène:
    115817
  • Synonymes antigène:
    Dehydrogenase/reductase SDR family member 1|dehydrogenase/reductase SDR family member 1|FLJ25430|DHRS1_HUMAN|MGC20204
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    34 kDa
  • Température de stockage:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Température de transport:
    4 °C
  • Immunogène:
    KLH conjugated synthetic peptide derived from human DHPS1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Type de conditionnement:
    Vial
  • Cdt:
    100 µl

Spécifications

À propos de cet article

DHRS1 (dehydrogenase/reductase (SDR family) member 1), also known as SDR19C1, is a 313 amino acid protein that belongs to the short-chain dehydrogenases/reductases (SDR) family and likely functions as an oxidoreductase. Abundantly expressed in heart and liver, DHRS1 contains an SDR motif and is encoded by a gene that maps to human chromosome 14q12. Human chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder ?-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.

Type: Primary
Antigen: DHPS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/313
Host: Rabbit
Isotype: IgG
Reactivity: Mouse