Order Entry
Belgium
ContactUsLinkComponent
Anti-GSK3B Rabbit Polyclonal Antibody
Anti-GSK3B Rabbit Polyclonal Antibody
  MSPP22104-1-AP
undefined
Anti-GSK3B Rabbit Polyclonal Antibody
  MSPP22104-1-AP

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Glycogen Synthase Kinase 3 beta
  • Symbole de l'antigène:
    GSK3B
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Synonymes antigène:
    EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
  • Storage buffer:
    PBS with 0,02% sodium azide and 50% glycerol pH 7,3.
  • Température de transport:
    Store at –20°C
  • Immunogène:
    Fusion Protein
  • Purification:
    Antigen affinity Purification
  • Cdt:
    20 µl

 

 

Glycogen synthase kinase-3 (GSK3) is a proline-directed serine-threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase .GSK3B is involved in energy metabolism, neuronal cell development, and body pattern formation.In skeletal muscle, it contributes to insulin regulation of glycogen synthesis by phosphorylating and inhibiting GYS1 activity and hence glycogen synthesis. Researches showed that the crystal structure of human GSK3B, expressed in insect cells, at 2,8-angstrom resolution .This antibody recognizse the C-terminal of GSK3B.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat