Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Catalog # COCO20250-20
Supplier: CONTROL
Specifications
- Plage de mesure:−200...1372 °C
- Résolution:0,1 °C; 1 °C (above 999,9 °C)
- Data memory - No. of readings:10000 data points
- Désignation:Pro 4-input data logging thermocouple thermometer, Type K, NIST-traceable calibration, 4 probe channels
- Sortie:USB
- Piles:9 V battery
- Affichage:Backlit LCD
- Précision:±0,15% of reading 1,0 °C, ±0,5% of reading 2,0 °C below –100 °C (±0,15% of reading 1,8 °F, ±0,5% of reading 3,6 °F below –148 °F)
- Cdt:1
Specifications
About this item
Rugged, heavy-duty construction protects against knocks and jars.
- Reliable measurements, guaranteed!
- Precalibrated to NIST-traceable standards
- Relative time stamp provides a time reference for major events
- Electronic off set function allows compensation of probe errors
- Infrared input port allows safe, noncontact measurements with optional IR wand
These professional type K thermometers provide accurate and reliable temperature measurement in an ergonomic design. Double-molded heavy-duty housing provides superior protection, making them ideal for tough applications. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
All models feature Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic offset function, temperature differential, backlit display, and auto power-off. Accept a wide variety of type K thermocouple probes with miniconnector.
Ordering information: These meters also feature an infrared input that accepts an optional IR wand for noncontact measurements of objects that are difficult or dangerous to reach; order wand 20250-24 separately. The IR temperature reading can be used as a unique input when calculating temperature differential. The IR wand has a distance-to sight ratio of 8:1 and temperature range of –35 to 550 °C. For more information; contact your Avantor representative.
Delivery information: Four Type K bead wire temperature probes, hard carrying case, USB cable, software, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.