Value Plastics® Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
N° de catalogue MFLX45508-30BLK
Fournisseur: Avantor Fluid Handling
Spécifications
- Taille de l'ardillon/Tube Ø int. (port 1):1/16"
- Type de connection:Female Luer to Barb
- Flow pattern:Straight
- Désignation:Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
- Seq:02
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- Cdt:1.000
- Fitting material:Polypropylene
Spécifications
À propos de cet article
Ensure your process integrity with full material traceability.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Taille de l'ardillon/Tube Ø int. (port 1):1/16"
- Type de connection:Female Luer to Barb
- Flow pattern:Straight
- Désignation:Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16"; 1000/PK
- Seq:02
- Environmentally Preferable:
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- Cdt:1.000
- Fitting material:Polypropylene