Smiths Medical® Fitting, Polycarbonate, Straight, Female Luer x Male Luer Lock, Smart Site; 1000/PK
N° de catalogue MFLX30526-10BLK
Fournisseur: Avantor Fluid Handling
Spécifications
- Type de connection:Female Luer to Male Luer
- Flow pattern:Straight
- Désignation:Fitting, Polycarbonate, Straight, Female Luer x Male Luer Lock, Smart Site; 1000/PK
- Seq:01
- Connection type (port 1):Female luer
- Connection type (port 2):Male luer lock
- No. of ports:2
- Garantie:Out of box
- Cdt:1.000
- Fitting material:Polycarbonate
Spécifications
À propos de cet article
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection type: Female Luer to Male Luer Lock with Smart Site Connection
Easily complete your luer fittings systems with these premium connectors. Included are some special configurations available only in polycarbonate. All female ports may be used with either slip or luer lock connections. Plugs and caps are also available. Polycarbonate material has good chemical compatibility and can be sterilized via ethylene oxide or gamma irradiation.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
- Type de connection:Female Luer to Male Luer
- Flow pattern:Straight
- Désignation:Fitting, Polycarbonate, Straight, Female Luer x Male Luer Lock, Smart Site; 1000/PK
- Seq:01
- Environmentally Preferable:
- Connection type (port 1):Female luer
- Connection type (port 2):Male luer lock
- No. of ports:2
- Garantie:Out of box
- Cdt:1.000
- Fitting material:Polycarbonate