Garmet hangers
Catalog # TEKMCRGS-GHO
Supplier: TEKNOMEK
Specifications
- Désignation:Garment hangers, open hook, 385W×200H mm
- Seq:02
- Cdt:1
Specifications
About this item
Open hook garment hangers are manufactured from 304 grade stainless steel in a polished finish.
- Open or closed hook garment hanger
- Fully welded 304 grade stainless steel
- Polished finish
Our open garment hangers are manufactured from fully welded 304 grade stainless steel with a polished finish. These are an ideal addition to our garment storage cupboards and garment rails.