Dey-Engley neutralising broth base dehydrated culture media
HIMEM187-500G
: HIMEDIA
:
- Cdt:500 g
- Désignation:Dey-Engley neutralizing broth base
Used along with Dey-Engley neutralising broth base for neutralizing and determining bactericidal activity of quaternary ammonium compounds.
- Microbiological culture media
- 500 g poly bottles