Ochratoxin A-solution 10 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Catalog # HONCR1964-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Type de conditionnement:Glass Bottle
- Cdt:2 mL
- Normes:Analytical standard
- Désignation:Ochratoxin A-solution 10 μg/mL in acetonitrile
- Conditions de stockage:-20°C
- MDL No.:MFCD00078079
- UN No.:1648
- Synonyme(s):(2S)-2-[[(3R)-5-chloro-8-hydroxy-3-methyl-1-oxo-3,4-dihydroisochromene-7-carbonyl]amino]-3-phenylpropanoic acid
- Formula:C₂₀H₁₈ClNO₆
- Concentration:10 μg/mL in acetonitrile
- Désignation:Ochratoxin A
- Point d'éclair:5 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery