Aflatoxin G2-solution 0.5 μg/mL in acetonitrile, Analytical standard, reference material, Fluka™
Catalog # HONCR1959-2ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Type de conditionnement:Glass Bottle
- Cdt:2 mL
- Normes:Analytical standard
- Désignation:Aflatoxin G2-solution 0.5 μg/mL in acetonitrile
- Conditions de stockage:-20°C
- MDL No.:MFCD00078141
- UN No.:1648
- Synonyme(s):(3S,7R)-11-methoxy-6,8,16,20-tetraoxapentacyclo[10.8.0.02,9.03,7.013,18]icosa-1,9,11,13(18)-tetraene-17,19-dione
- Formula:C₁₇H₁₄O₇
- Concentration:0.5 μg/mL in acetonitrile
- Désignation:Aflatoxin G2
- Point d'éclair:5 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery