Order Entry
Belgium
ContactUsLinkComponent
 
1-Hexanol Analytical standard, reference material, Fluka™
  HONCR1931-1G
 :  
1-Hexanol Analytical standard, reference material, Fluka™
  HONCR1931-1G
 :  

 

  • Type de conditionnement:
    Glass Bottle
  • Cdt:
    1 g
  • Normes:
    Analytical standard
  • N ° CAS.:
    111-27-3
  • Désignation:
    1-Hexanol
  • Conditions de stockage:
    RT
  • MDL No.:
    MFCD00002982
  • UN No.:
    2282
  • Synonyme(s):
    Hexyl alcohol
  • Point de fusion:
    -52 °C
  • Formula:
    C₆H₁₄O
  • Molecular weight:
    102.18 g/mol
  • Désignation:
    1-Hexanol
  • Point d'éclair:
    59 °C
  • Physical form:
    Liquid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery