Fluoxastrobin Analytical standard, reference material, Fluka™
HONCR1893-100MG
:
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:361377-29-9
- Désignation:Fluoxastrobin
- Conditions de stockage:RT
- MDL No.:MFCD07363979
- Synonyme(s):(1E)-[2-[[6-(2-Chlorophenoxy)-5-fluoro-4-pyrimidinyl]oxy]phenyl](5,6-dihydro-1,4,2-dioxazin-3-yl)methanone O-methyloxime
- Point de fusion:103 - 108 °C
- Formula:C₂₁H₁₆ClFN₄O₅
- Molecular weight:458.83 g/mol
- Désignation:Fluoxastrobin
- Physical form:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery