beta-HCH Analytical standard, reference material, Fluka™
Catalog # HONCR1884-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:319-85-7
- Désignation:beta-HCH
- Conditions de stockage:RT
- MDL No.:MFCD00135948
- UN No.:2811
- Synonyme(s):ß-1,2,3,4,5,6-Hexachlorocyclohexane
- Point de fusion:309 °C
- Formula:C₆H₆Cl₆
- Molecular weight:290.83 g/mol
- Désignation:beta-HCH
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery