Emamectine benzoate Analytical standard, reference material, Fluka™
Catalog # HONCR1870-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:155569-91-8
- Désignation:Emamectine benzoate
- Conditions de stockage:RT
- MDL No.:MFCD01757274
- UN No.:2811
- Synonyme(s):(4′’R)-4″-Deoxy-4″-(methylamino)avermectin B1 benzoate
- Point de fusion:152 °C
- Formula:C₅₆H₈₁NO₁₅
- Molecular weight:1008.2 g/mol
- Désignation:Emamectine benzoate
- Physical form:powder
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery