Order Entry
Belgium
ContactUsLinkComponent
Ampicillin trihydrate Analytical standard, reference material, Fluka™
Ampicillin trihydrate Analytical standard, reference material, Fluka™
Catalog # HONCR1838-100MG
CAS Number:  
Ampicillin trihydrate Analytical standard, reference material, Fluka™
Catalog # HONCR1838-100MG
CAS Number:  

Specifications

  • Type de conditionnement:
    Glass Bottle
  • Cdt:
    100 mg
  • Normes:
    Analytical standard
  • N ° CAS.:
    7177-48-2
  • Désignation:
    Ampicillin trihydrate
  • Conditions de stockage:
    2-8°C
  • MDL No.:
    MFCD00072036
  • Synonyme(s):
    (2S,5R,6R)-6-{[(2R)-2-Amino-2-phenylacetyl]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
  • Point de fusion:
    195 - 200 °C
  • Formula:
    C₁₆H₁₉N₃O₄S·₃H₂O
  • Molecular weight:
    403.45 g/mol
  • Désignation:
    Ampicillin trihydrate
  • Physical form:
    Crystalline

Specifications

About this item

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery