Tolclofos-methyl Analytical standard, reference material, Fluka™
HONCR1832-100MG
:
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:57018-04-9
- Désignation:Tolclofos-methyl
- Conditions de stockage:RT
- MDL No.:MFCD00078741
- Synonyme(s):O-(2,6-Dichloro-4-methylphenyl) O,O-dimethyl phosphorothioate
- Point de fusion:79 - 79.5 °C
- Formula:C₉H₁₁Cl₂O₃PS
- Molecular weight:301.1 g/mol
- Désignation:Tolclofos-methyl
- Physical form:Solid
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery