Order Entry
Belgium
ContactUsLinkComponent
 
Methidathion Analytical standard, reference material, Fluka™
  HONCR1813-100MG
 :  
Methidathion Analytical standard, reference material, Fluka™
  HONCR1813-100MG
 :  

 

  • Type de conditionnement:
    Glass Bottle
  • Cdt:
    100 mg
  • Normes:
    Analytical standard
  • N ° CAS.:
    950-37-8
  • Désignation:
    Methidathion
  • Conditions de stockage:
    2-8°C
  • MDL No.:
    MFCD00055286
  • UN No.:
    2811
  • Synonyme(s):
    S-2,3-Dihydro-5-methoxy-2-oxo-1,3,4-thiodiazol-3-yl-methyl-O,O-dimethyl-phosphodithioate
  • Point de fusion:
    39 - 40 °C
  • Formula:
    C₆H₁₁N₂O₄PS₃
  • Molecular weight:
    302.3 g/mol
  • Désignation:
    Methidathion
  • Physical form:
    Solid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery