Chlorosulphuron Analytical standard, reference material, Fluka™
Catalog # HONCR1812-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:64902-72-3
- Désignation:Chlorosulphuron
- Conditions de stockage:Ambient
- MDL No.:MFCD00128059
- Synonyme(s):1-(2-Chlorophenylsulfonyl)-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea
- Formula:C₁₂H₁₂ClN₅O₄S
- Désignation:Chlorosulphuron
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery