Parathion-methyll Analytical standard, reference material, Fluka™
N° de catalogue HONCR1800-100MG
Fournisseur: Honeywell Chemicals
Numéro CAS:
Spécifications
- Type de conditionnement:Glass Bottle
- Cdt:100 mg
- Normes:Analytical standard
- N ° CAS.:298-00-0
- Désignation:Parathion-methyll
- Conditions de stockage:2-8°C
- MDL No.:MFCD00041813
- UN No.:2811
- Point de fusion:36 °C
- Molecular weight:263.21 g/mol
- Désignation:Parathion-methyll
- Physical form:Crystalline
Spécifications
À propos de cet article
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery