Indicator, Click-On In-Line Super Clean®, Restek
Catalog # REST22474
Supplier: Restek
Specifications
- Désignation:Click-on in-line super clean indicator (oxygen, moisture)
- Cdt:1
Specifications
About this item
Install an indicator after the Click-On in-line gas filter so there is no confusion about when to replace the traps.
- Colored indicator for detecting buildup of contaminants
- Oxygen: green to gray
- Moisture: beige to clear
Ordering information: Note: Fittings sold separately.