Microplate incubator shaker, Jitterbug™
Catalog # BOEI130000
Supplier: BOEKEL SCIENTIFIC
Specifications
- Orbit:1 mm
- Gamme de vitesse:575 - 1500 min⁻¹
- Désignation:Microplate incubator shaker
- Cdt:1
Specifications
About this item
Quiet, rugged shaker is ideal for a variety of immunoassay and research applications.
Accommodates two microplates. Microprocessor-based for independent setting of temperature, shaking speed, and mixing duration. The warmed inner aluminium bed provides temperature uniformity. An inner chamber cover reduces the risk of biohazard from the disbursement of aerosols and insulates the plates during incubation. Outer tinted Plexiglas® cover further insulates and protects sensitive reagents from light. A circulating fan with auto shutoff prevents condensation. Timed operation from 1 second to 99 min., 59 seconds (with 1 second resolution) or unit may be “pulsed” for three seconds. Eight selectable speeds. ABS plastic with a metal base.
Certifications: UL listed.