Pharmalabels 'Sampled'
Pharmalabels 'Sampled'
Supplier:   Sampling Systems
Pharmalabels 'Sampled'
Supplier:  Sampling Systems
Certificates
Pharmalabels 'Sampled'
Pharmalabels 'Sampled'
2 Options Available Below
Product Details & Documents
Pre-printed 'Sampled' labels feature a write-on paper front.
  • Seals holes in bags
  • The pharma range have FDA ccompliant glue
Pre-printed "Sampled" labels feature a write-on paper front and a high strength, ultra adhesive. The UA range have a foil backing to provide a stronger barrier with the product. The pharma range have an FDA compliant glue. The high strength adhesive used on these labels allows them to stick on a wide range of surfaces, including very dusty areas. The working temperature range is −20 to 70 °C (−5 to 160 °F).
Product Family Options
Product Information
  • Largeur
    Longueur
    Emballage
    Availability
    Price
  • 95 mm
    95 mm
    500/roll
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
  • 150 mm
    150 mm
    250/roll
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €