Masterflex® Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 25/PK
N° de catalogue MFLX45509-04
Fournisseur: Avantor Fluid Handling
Avantor exclusive
Spécifications
- Type de connection:Threaded Nut
- Flow pattern:Straight
- Désignation:Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 25/PK
- Garantie:3 Months
- Cdt:25
- Fitting material:Nylon
Spécifications
À propos de cet article
Best for use with panel mount fittings.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Optimise your application with these luer fitting accessories.
- Type de connection:Threaded Nut
- Flow pattern:Straight
- Désignation:Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 25/PK
- Environmentally Preferable:
- Garantie:3 Months
- Cdt:25
- Fitting material:Nylon