Masterflex® Electro-Optic Optical Sensor Controller, Avantor®
Optical Sensor Controller; Allows For Single-point Detection And Control, 585 mm, 7,62 cm, 12,5 cm, 120 VAC
N° de catalogue MFLX32772-50
Fournisseur: Avantor Fluid Handling
Masterflex® Electro-Optic Optical Sensor Controller, Avantor®
N° de catalogue MFLX32772-50
Fournisseur: Avantor Fluid Handling
Aller à :
Product Details & Documents
Converts standard 110 VAC line current to 10 to 28 VDC input power.
- Easily covert VAC to VDC
Controller converts standard 110 VAC line current to the 10 to 28 VDC input power required for optical sensor operation, and provides an SPDT, 5 A relay output for direct control of moderate loads. Input voltage: 120 VAC, 50/60Hz. Max current draw: 70 mA at 120 VAC. Operating temperature: –13 to 158 °F (–25 to 70 °C). Electrical connections: eight ¼" male spade terminals. Dimensions: 3"L×23-1/32"W×4-15/16"H.
Spécifications
- Largeur:585 mm
- Désignation:Optical Sensor Controller; Allows For Single-point Detection And Control
- Hauteur:12,5 cm
- Longueur:7,62 cm
- Puissance:120 VAC
- Cat. No.:MFLX32772-50
- Min. process temperature:-13 °F
- Max. process temperature:70 °C
- Garantie:1 year