Masterflex® L/S® Standard Pump Head Replacement Thrust Washers; 10/PK
N° de catalogue MFLX07021-04
Fournisseur: Avantor Fluid Handling
Spécifications
- Pump series:L/S
- Désignation:Replacement thrust washers for L/S Standard pump heads, pack of 10,
- Environmentally Preferable:
- Cdt:10
Spécifications
À propos de cet article
Get the most out of your Masterflex® L/S® Standard Pump Head with these replacement parts.
- Suitable for use with Masterflex® L/S® Standard pump heads
Replacement parts to keep your pump head operating even longer. Thrust washers fit on either side of the rotor assembly of L/S standard pump heads.
- Pump series:L/S
- Désignation:Replacement thrust washers for L/S Standard pump heads, pack of 10,
- Environmentally Preferable:
- Cdt:10