Masterflex® Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, 3/4" Mini x 1/2" NPT(F)
Catalog # MFLX31200-13
Supplier: Avantor Fluid Handling
CE certificate
Specifications
- Type de connection:Sanitary to Thread
- Flow pattern:Straight
- Sanitary clamp size (Port 1):¾"
- Thread size NPT(F) (port 2):½"
- Désignation:Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, ¾" x ½" NPT(F)
- Seq:04
- Cdt:1
- Fitting material:Polypropylene
- Sanitary gasket size:¾"
Specifications
About this item
Maintain sanitary conditions.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most materials and configurations available to ship same day
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
- All fittings are compatible with Tri-Clamp® connections
- Excellent chemical resistance; low leachable characteristics
- Connection Type: 3/4" mini to 1/2" NPT(F)
These fittings are ideal for use with the Masterflex L/S tubing pumps.
- Type de connection:Sanitary to Thread
- Flow pattern:Straight
- Sanitary clamp size (Port 1):¾"
- Thread size NPT(F) (port 2):½"
- Désignation:Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, ¾" x ½" NPT(F)
- Seq:04
- Environmentally Preferable:
- Cdt:1
- Fitting material:Polypropylene
- Sanitary gasket size:¾"