ContactUsLinkComponent
Masterflex® C/L® Pump Power Supply, Avantor®
Masterflex® C/L® Power Supply for VDC Powered Variable-Speed Pump; 100 to 240 VAC
N° de catalogue MFLX77120-11
Fournisseur:  Avantor Fluid Handling
CE Compliant
undefined
Masterflex® C/L® Power Supply for VDC Powered Variable-Speed Pump; 100 to 240 VAC
N° de catalogue MFLX77120-11
Fournisseur:  Avantor Fluid Handling
CE Compliant
Spécifications
  • Désignation:
    Accessory Replacement power supply, 115/230 VAC
  • Garantie:
    1 year
  • Cdt:
    1
Spécifications
À propos de cet article
Safely adapt your VDC powered C/L® pump.
  • Suitable for use with C/L® pumps
Easily convert your 12 VDC C/L® Variable-Speed pumps to 100 to 240 VAC power. This power supply is suitable for use with 12 VDC models 77122-06, -16, and 26 single-channel pumps and models 77120-52 and -62 dual-channel pumps.
  • Désignation:
    Accessory Replacement power supply, 115/230 VAC
  • Environmentally Preferable:
  • Garantie:
    1 year
  • Cdt:
    1