Dumont #5, ceramic coated forceps
#5, Inox, ceramic coated, 110 mm, Biology, Straight, 0,05×0,01 mm
N° de catalogue FSCI11252-50
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Dumont #5, ceramic coated forceps
N° de catalogue FSCI11252-50
Fournisseur: FINE SCIENCE TOOLS
Marketsource
Aller à :
Product Details & Documents
Spécifications
- Type:#5
- Longueur:110 mm
- Matière:Inox, ceramic coated
- Style de pointe:Biology
- Tip size:0,05×0,01 mm
- Tip shape:Straight