Fine forceps, broad shanks
Fine, Acier inoxydable, 120 mm, Broad shanks, Straight, 0,2×0,12 mm
Catalog # FSCI11445-12
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
Specifications
- Type:Fine
- Longueur:120 mm
- Matière:Acier inoxydable
- Style de pointe:Broad shanks
- Tip size:0,2×0,12 mm
- Tip shape:Straight