Titanium micro serrefines
Marketsource
Supplier:   FINE SCIENCE TOOLS
Titanium micro serrefines
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Titanium micro serrefines
Titanium micro serrefines
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Style de pointe
    Tip shape
    Tip size
    Availability
    Price
  • Strong clamp press, 125 g
    Titane
    13 mm
    Serrated
    Straight
    4×0,75 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 133,00 €
    133,00 €
    1 * 1 Pce
       133,00 €
    Specifications:
    Marketsource
    Marketsource
  • Medium clamp press, 100 g
    Titane
    15 mm
    Serrated
    Straight
    6×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 133,00 €
    133,00 €
    1 * 1 Pce
       133,00 €
    Specifications:
    Marketsource
    Marketsource
  • Light clamp press, 85 g
    Titane
    16 mm
    Serrated
    Straight
    8×2 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 133,00 €
    133,00 €
    1 * 1 Pce
       133,00 €
    Specifications:
    Marketsource
    Marketsource
  • Light clamp press, 60 g
    Titane
    19 mm
    Serrated
    Straight
    10×2 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 133,00 €
    133,00 €
    1 * 1 Pce
       133,00 €
    Specifications:
    Marketsource
    Marketsource