Surgical scissors, large loops
Sharp/blunt, straight tips, large loops, 145 mm, 45 mm
N° de catalogue FSCI14101-14
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Surgical scissors, large loops
N° de catalogue FSCI14101-14
Fournisseur: FINE SCIENCE TOOLS
Marketsource
Aller à :
Product Details & Documents
These scissors are made of stainless steel.
Spécifications
- Type:Sharp/blunt, straight tips, large loops
- Longueur:145 mm
- Longueur de lame:45 mm