Tissue separating scissors
Blunt/blunt, straight tips, 115 mm, 23 mm
N° de catalogue FSCI14072-10
Marketsource
Fournisseur: FINE SCIENCE TOOLS
Aller à :
Product Details & Documents
These scissors are made of stainless steel.
Spécifications
- Type:Blunt/blunt, straight tips
- Longueur:115 mm
- Longueur de lame:23 mm