Scissors with tweezer tips, Rubis®
N° de catalogue 233-0226
Fournisseur: OUTILS RUBIS
Spécifications
- Type:Straight, tweezers tips
- Longueur:90 mm
- Environmentally Preferable:
- Cdt:1
- Tip size:5 mm
Spécifications
À propos de cet article
Allround tweezers engineered into a scissors-movement.
- Produced from hardened stainless steel AISI420
- Highly resistant against acids and alkaline
- With ergonomically designed Rubis Twist® grips and high precison tweezers tips
- No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.