Anti-SOD1 Mouse Monoclonal Antibody [clone: O98B10]
- Catalog No:
- BLEG850702
- BLEG850701
- Symbole de l'antigène:
- SOD1
- SOD1
- Synonymes antigène:
- Cystolic|Superoxide dismutase 1|Superoxide dismutase [Cu-Zn] |Superoxide Dismutase
- Cystolic|Superoxide dismutase 1|Superoxide dismutase [Cu-Zn] |Superoxide Dismutase
- Clonalité:
- Monoclonal
- Monoclonal
- Clone:
- O98B10
- O98B10
- Réactivité:
- Drosophila, Human, Rat, Mouse
- Drosophila, Human, Rat, Mouse
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- Immunogène:
- Full-length human SOD1 expressed in 293T cells transfected with human SOD1 expression vector.
- Full-length human SOD1 expressed in 293T cells transfected with human SOD1 expression vector.
- Format:
- Purified
- Purified
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.5 mg/ml
- 0.5 mg/ml
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Température de stockage:
- 2…8 °C
- 2…8 °C
- Tested applications:
- WB
- WB
Frequently Bought Together