Test strips, QUANTOFIX Total Chlorine Sensitive 1
Catalog # 634-1190
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Cdt:50
- Désignation:QUANTOFIX Total Chlorine Sensitive 1, 50 tests
Specifications
About this item
Semi-quantitative test strips for reliable determination of total chlorine concentrations as low as 0,01 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad, providing a high level of sensitivity. Ideal for rapid and easy analysis of total chlorine directly on-site. No maintenance or calibration is required.
- Sensitive rapid test