Lambda gt11 vector kits
N° de catalogue STRA234211
Fournisseur: STRATAGENE
Numéro CAS:
Spécifications
- Cdt:1
- Désignation:Lambda gt11/EcoR I/CIAP Treated Vector Kit, vector arms dephosphorylated with alkaline phosphatase to prevent self-ligation. Contains: 10 μg lambda gt11/EcoR I/CIAP-treated vector, host strains: Y1088, Y1089r-, Y1090r-, test insert
- Seq:01
Spécifications
À propos de cet article
For use in the construction of cDNA libraries.
- Predigested with EcoR I
- >1×10⁷ pfu/μg lambda DNA must be obtained with test insert (background <1,5%)
These Lambda gt11 vector kits do not allow in vivo excision of cloned fragments. Libraries may be screened with both antibodies and nucleic acid probes.
Attention: For research use only. Not for use in diagnostic procedures.