HRP-conjugate stabiliser, SeramunStab® STA2
Catalogus # APOSBIL3112-100ML
Leverancier: Apollo Scientific
CAS-nummer:
Specificaties
- Cdt:100 mL
- Hôte:
- Désignation:SeramunStab® STA2 HRP-Conjugate stabiliser
- Seq:01
- Environmentally Preferable:
Specificaties
Over dit artikel
Liquid product red odourless pH-value 7,3 to 7,5 Boiling point 102 °C. Stabiliser for a broad range of HRP conjugates.