Specifications
- Désignation:Filter gas charcoal/13X
- Cdt:1
Specifications
About this item
These gas purifying cartridges for ICP-OES are a cost effective safety and preventative measure to assure the supply of the highest purity gas to the ICP-OES instrumentation.
- Deliver clean gases for accurate analyzes
- Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream
- Easily replaced
Impure gases can cause installation delays, premature instrument failure, and flawed results. Purification is one of the most important steps you can take to optimize your ICP-OES system performance. Agilent brings the highest performance of gas purifiers to ICP-OES instrument users. We manufacture our own range of gas purifying cartridges specifically designed to remove impurity gases, moisture and hydrocarbons from the ICP-OES gas supplies. We recommend that each Agilent ICP-OES system uses a gas purification system with Agilent gas purifying cartridges. These systems ensure the highest quality gas and include a tag to remind you when replacement is required, so you can replace them before sensitivity decreases, results are impacted or the instrument suffers other failures.