Proteinase K
Proteinase K
1
N° de catalogue AGLS300140
Fournisseur:   AGILENT
Proteinase K
N° de catalogue AGLS300140
Fournisseur:  AGILENT
Numéro CAS:  
Proteinase K
1
Numéro CAS:  
Prix
223,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Proteinase K is a broad spectrum protease from Tritirachium album that is ideal for preparing chromosomal DNA.
  • For general digestion of protein in biological samples
  • Lyophilised powder with specific activity >30 units/mg at 37 °C
  • Size: 100 mg
It cleaves peptide bonds in which carbonyl groups are surrounded by aromatic, hydrophobic, or aliphatic amino acids.
Used for the following applications: pulsed field electrophoresis, chromosomal DNA for protein fingerprinting, and nuclease removal from DNA and RNA preparations.
Attention: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
  • Cdt:
    1
  • Cat. No.:
    AGLS300140
  • Nom de l'enzyme:
    Proteinase K
Spécifications