SCSI supercompetent cells
SCSI supercompetent cells
SCSI supercompetent cells, size: 5×0,2 ml; 1×10⁹ transformants/µg
N° de catalogue AGLS200231
Fournisseur:   AGILENT
SCSI supercompetent cells
N° de catalogue AGLS200231
Fournisseur:  AGILENT
SCSI supercompetent cells
SCSI supercompetent cells, size: 5×0,2 ml; 1×10⁹ transformants/µg
Prix
315,00 €
Connexion Pour la disponibilité
1 * 1 mL
Product Details & Documents
SCSI supercompetent and DH5α competent cells are classic E. coli strains engineered for competence.
  • Engineered to become competent
This collection of competent cells, including SCSI supercompetent cells and DH5α competent cells, consists of classic E. coli strains that have been engineered to become competent for use in cloning experiments.
Spécifications
  • Désignation:
    SCSI supercompetent cells, size: 5×0,2 ml; 1×10⁹ transformants/µg