Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Protéines et Peptides
Recombinant Hepatitis C Virus NS4 a+b, Rhodamine
Recombinant Hepatitis C Virus NS4 a+b, Rhodamine
Recombinant Hepatitis C Virus NS4 a+b, Rhodamine
APOSBITP1793-1MG
:
Apollo Scientific
:
Recombinant Hepatitis C Virus NS4 a+b, Rhodamine
APOSBITP1793-1MG
:
Apollo Scientific
:
Cdt:
1 mg
Conjugaison:
Rhodamine
Type de protéine/peptide:
Recombinant
Synonyme de protéine:
HCV NS4 a+b
Nom de la protéine/du peptide:
Hepatitis C Virus NS4 a+b