Order Entry
Belgium
ContactUsLinkComponent
Methyl-3-(chlorosulphonyl)-4-methoxybenzoate 95%
Methyl-3-(chlorosulphonyl)-4-methoxybenzoate 95%
Catalog # APOSOR42015-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3-(chlorosulphonyl)-4-methoxybenzoate 95%
Catalog # APOSOR42015-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Type de conditionnement:
    Bottle
  • Cdt:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.69 g/mol
Melting Pt:126…128 °C
Storage Temperature:Température ambiante
UN:3261
ADR:8,III
 :GHS05
CAS Number:192323-12-9
Formula:C₉H₉ClO₅S
MDL Number:MFCD03407355